4-Amino-6-chloropyrimidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
6-Chloropyrimidin-4-ylamine, 6-Chloropyrimidin-4-amine, 6-Chloro-4-pyrimidinamine, 6-Chlor-4-pyrimidinamin
A2136-1G
A2136-5G
TCIAA2136-1GEA
31
EUR
TCIAA2136-1G
TCIAA2136-5G
4-Amino-6-chloropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-6-chloropyrimidine
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...