Stampa…

6-Aminouracil ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 6-Amino-2,4-dihydroxypyrimidine, 6-Aminopyrimidine-2,4(1H,3H)-dione, 4-Amino-2,6-dihydroxypyrimidine, 4-Aminouracil

A0658-25G A0658-250G
TCIAA0658-25GEA 43 EUR
TCIAA0658-25G TCIAA0658-250G
6-Aminouracil ≥98.0% (by HPLC, titration analysis)
6-Aminouracil
Formula: C₄H₅N₃O₂
Peso molecolare: 127,10 g/mol
Punto di fusione: 360 °C
MDL Number: MFCD00006071
CAS Number: 873-83-6
EINECS: 212-854-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45f43f7b2b4ba7489816595b17dd179e2ad91d0c&vid=45f43f7b2b4ba7489816595b17dd179e2ad91d0c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR