Stampa…
4,6-Dihydroxypyrimidine ≥95.0% (by titrimetric analysis)
Fornitore: TCI
Attenzione
D1821-25G D1821-250G
TCIAD1821-25GEA 30 EUR
TCIAD1821-25G TCIAD1821-250G
4,6-Dihydroxypyrimidine ≥95.0% (by titrimetric analysis)
4,6-Dihydroxypyrimidine
Formula: C₄H₄N₂O₂
Punto di fusione: > 300 °C
MDL Number: MFCD00016733
CAS Number: 1193-24-4
EINECS: 214-772-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f71e2150c78cda1d1df7dbe5730a5c71ebab531&vid=6f71e2150c78cda1d1df7dbe5730a5c71ebab531&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 95.0 %