α-Bromo-p-toluic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Bromomethylbenzoic acid, 4-(Bromomethyl)benzoic acid
B0845-25G
B0845-5G
TCIAB0845-25GEA
115
EUR
TCIAB0845-25G
TCIAB0845-5G
α-Bromo-p-toluic acid ≥97.0% (by GC, titration analysis)
α-Bromo-p-toluic acid
Formula:
C₈H₇BrO₂ Peso molecolare: 215,05 g/mol Punto di ebollizione: 316 °C (1013 hPa) Punto di fusione: 223…226 °C |
MDL Number:
MFCD00002567 CAS Number: 6232-88-8 EINECS: 228-343-3 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...