Stampa…

3-Thenoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

T1084-25G T1084-5G
TCIAT1084-25GEA 147 EUR
TCIAT1084-25G TCIAT1084-5G
3-Thenoic acid ≥98.0% (by GC, titration analysis)
3-Thenoic acid
Formula: C₅H₄O₂S
Peso molecolare: 128,15 g/mol
Punto di fusione: 137 °C
MDL Number: MFCD00005467
CAS Number: 88-13-1
EINECS: 201-802-5
Merck Index: 14,09279
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feee71a15c5409f32e2e0ce92e52c2328a4ddbf5&vid=feee71a15c5409f32e2e0ce92e52c2328a4ddbf5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR