Stampa…

3-Nitrophthalimide ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

N0666-25G N0666-5G
TCIAN0666-25GEA 190 EUR
TCIAN0666-25G TCIAN0666-5G
3-Nitrophthalimide ≥97.0% (by titrimetric analysis)
3-Nitrophthalimide
Formula: C₈H₄N₂O₄
Peso molecolare: 192,13 g/mol
Punto di fusione: 218 °C
MDL Number: MFCD00041852
CAS Number: 603-62-3
EINECS: 210-051-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e13d31457826d3e5729038ef795fdfe4b8ecdac&vid=3e13d31457826d3e5729038ef795fdfe4b8ecdac&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR