Stampa…

3-Nitrophthalic anhydride ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

N0245-25G N0245-250G
TCIAN0245-25GEA 34 EUR
TCIAN0245-25G TCIAN0245-250G
3-Nitrophthalic anhydride ≥97.0% (by GC, titration analysis)
3-Nitrophthalic anhydride
Formula: C₈H₃NO₅
Peso molecolare: 193.12 g/mol
MDL Number: MFCD00005921
CAS Number: 641-70-3
EINECS: 211-373-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43bdc4d6ee6548460bf6b9a617df8bfb2b2d51e0&vid=43bdc4d6ee6548460bf6b9a617df8bfb2b2d51e0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Methanolysis Method) min. 97.0 %
Melting point 161.0 to 165.0 deg-C
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR