Butyl isovalerate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Isovaleric acid butyl ester, Butyl 3-methylbutanoate
B3718-25ML
B3718-500ML
TCIAB3718-25MLEA
16
EUR
TCIAB3718-25ML
TCIAB3718-500ML
Butyl isovalerate ≥98.0% (by GC)
Butyl isovalerate
Formula:
C₉H₁₈O₂ Peso molecolare: 158,24 g/mol Punto di ebollizione: 176°C Flash Pt: 53°C |
MDL Number:
MFCD00048775 CAS Number: 109-19-3 EINECS: 203-654-7 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...