3-Amino-5-methylisoxazole ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Methylisoxazol-3-amine, 3-Amino-5-methylisoxazol, 5-Methylisoxazol-3-ylamine
A0980-25G
A0980-250G
TCIAA0980-25GEA
79
EUR
TCIAA0980-25G
TCIAA0980-250G
3-Amino-5-methylisoxazole ≥97.0% (by GC, titration analysis)
3-Amino-5-methylisoxazole
Formula:
C₄H₆N₂O Peso molecolare: 98,1 g/mol Punto di ebollizione: 194 °C (1013 hPa) Punto di fusione: 56…60 °C |
MDL Number:
MFCD00003155 CAS Number: 1072-67-9 EINECS: 214-013-6 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 57.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...