Stampa…

3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

D2381-25G D2381-250G
TCIAD2381-25GEA 46 EUR
TCIAD2381-25G TCIAD2381-250G
3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Di-tert-butyl-4-hydroxybenzoic acid
Formula: C₁₅H₂₂O₃
Punto di ebollizione: 374 °C (1013 hPa)
Punto di fusione: 219…220 °C
MDL Number: MFCD00008827
CAS Number: 1421-49-4
EINECS: 215-823-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 213.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR