Stampa…

Methyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate

M1943-25G M1943-5G
TCIAM1943-25GEA 75 EUR
TCIAM1943-25G TCIAM1943-5G
Methyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl-3,4-dihydroxybenzoate
Formula: C₈H₈O₄
Peso molecolare: 168,15 g/mol
MDL Number: MFCD00016437
CAS Number: 2150-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd0cb041201ecf2d41072ddf25d5463698d664e5&vid=fd0cb041201ecf2d41072ddf25d5463698d664e5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR