Stampa…
Ethyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate
D0571-25G D0571-5G
TCIAD0571-25GEA 55 EUR
TCIAD0571-25G TCIAD0571-5G
Ethyl-3,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Ethyl-3,4-dihydroxybenzoate
Formula: C₉H₁₀O₄
Peso molecolare: 182,18 g/mol
Punto di fusione: 134 °C
MDL Number: MFCD00002199
CAS Number: 3943-89-3
EINECS: 223-529-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9612e016a4e8bd69f6cf7b3ca4ced2c370b6b6ae&vid=9612e016a4e8bd69f6cf7b3ca4ced2c370b6b6ae&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 133.0 to 136.0 deg-C
Solubility in Methanol almost transparency