Stampa…

3,3,3-Triphenylpropionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

T1558-5G T1558-25G
TCIAT1558-5GEA 69 EUR
TCIAT1558-5G TCIAT1558-25G
3,3,3-Triphenylpropionic acid ≥98.0% (by GC, titration analysis)
3,3,3-Triphenylpropionic acid
Formula: C₂₁H₁₈O₂
Punto di fusione: 184 °C
Densità: 1,264 g/cm³ (20 °C)
MDL Number: MFCD00002713
CAS Number: 900-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=157ed551d9ec06abede7f24fc014e3f2b0048551&vid=157ed551d9ec06abede7f24fc014e3f2b0048551&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR