3-(3-Fluorophenyl)propionic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(3-Fluorophenyl)propanoic acid
F0779-1G
F0779-5G
TCIAF0779-1GEA
23
EUR
TCIAF0779-1G
TCIAF0779-5G
3-(3-Fluorophenyl)propionic acid ≥97.0% (by GC, titration analysis)
3-(3-Fluorophenyl)propionic acid
Formula:
C₉H₉FO₂ Peso molecolare: 168,17 g/mol Punto di ebollizione: 119°C |
MDL Number:
MFCD01310821 CAS Number: 458-45-7 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...