Isobutyric anhydride ≥95.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
2-Methylpropanoic anhydride, 2-Methylpropionic anhydride
Formula:
C₈H₁₄O₃ Peso molecolare: 158.2 g/mol Punto di ebollizione: 182°C Punto di fusione: -69 Flash Pt: 64°C |
MDL Number:
MFCD00008913 CAS Number: 97-72-3 EINECS: 202-603-6 UN: 3265 ADR: 8 III |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Morpholine Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...