Ethyl-O-toluate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl-2-methylbenzoate, 2-Toluic acid ethyl ester, o-Toluic acid ethyl ester, Ethyl 2-methylbenzoate, 2-Methylbenzoic acid ethyl ester
T0294-25G
T0294-500G
TCIAT0294-25GEA
28
EUR
TCIAT0294-25G
TCIAT0294-500G
Ethyl-O-toluate ≥98.0% (by GC)
Ethyl-O-toluate
Formula:
C₁₀H₁₂O₂ Peso molecolare: 164,2 g/mol Punto di ebollizione: 227°C Punto di fusione: –10 °C Densità: 1,0479 g/cm³ (4 °C) Flash Pt: 91°C |
MDL Number:
MFCD00009112 CAS Number: 87-24-1 EINECS: 201-734-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...