2-Methoxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
M2191-1G
M2191-5G
TCIAM2191-1GEA
21
EUR
TCIAM2191-1G
TCIAM2191-5G
2-Methoxy-3-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Methoxy-3-(trifluoromethyl)pyridine
Formula:
C₇H₆F₃NO Peso molecolare: 177,13 g/mol Flash Pt: 47°C |
MDL Number:
MFCD00153202 CAS Number: 121643-44-5 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...