Stampa…

2-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Carboxybenzaldehyde, Phthalaldehydic acid

P0281-25G P0281-500G P0281-100G
TCIAP0281-25GEA 34 EUR
TCIAP0281-25G TCIAP0281-500G TCIAP0281-100G
2-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Formylbenzoic acid
Formula: C₈H₆O₃
Peso molecolare: 150,13 g/mol
Punto di ebollizione: 101 °C (0,4 -rr)
Punto di fusione: 95…98 °C
Densità: 1,404 g/cm³ (20 °C)
MDL Number: MFCD00003336
CAS Number: 119-67-5
EINECS: 204-342-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fac90e63c4c1bcf859a00161896207e06afa80dc&vid=fac90e63c4c1bcf859a00161896207e06afa80dc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR