Stampa…

2-Fluoro-p-anisaldehyde ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Fluoro-4-methoxybenzaldehyde, 2-Fluoro-4-anisaldehyde

F0386-25G F0386-5G
TCIAF0386-25GEA 160 EUR
TCIAF0386-25G TCIAF0386-5G
2-Fluoro-p-anisaldehyde ≥96.0% (by GC)
2-Fluoro-p-anisaldehyde
Formula: C₈H₇FO₂
Peso molecolare: 154,14 g/mol
MDL Number: MFCD00236679
CAS Number: 331-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80d65b05934743c44a51e1cc8eae6fda6e02c417&vid=80d65b05934743c44a51e1cc8eae6fda6e02c417&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 45.0 to 48.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR