Stampa…

2-Chloroisonicotinonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Chloro-4-cyanopyridine, 2-Chloropyridine-4-carbonitrile

C1873-25G C1873-5G
TCIAC1873-25GEA 122 EUR
TCIAC1873-25G TCIAC1873-5G
2-Chloroisonicotinonitrile ≥98.0% (by GC)
2-Chloroisonicotinonitrile
Formula: C₆H₃ClN₂
Peso molecolare: 138,56 g/mol
Punto di ebollizione: 106°C
MDL Number: MFCD00174318
CAS Number: 33252-30-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb7ba552ab70600557ec3125c42ee4d141801700&vid=fb7ba552ab70600557ec3125c42ee4d141801700&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR