Stampa…

2-Amino-6-ethoxybenzothiazole ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 6-Ethoxybenzothiazol-2-ylamine

A0716-25G A0716-500G
TCIAA0716-25GEA 30 EUR
TCIAA0716-25G TCIAA0716-500G
2-Amino-6-ethoxybenzothiazole ≥97.0% (by titrimetric analysis)
2-Amino-6-ethoxybenzothiazole
Formula: C₉H₁₀N₂OS
Punto di ebollizione: 347 °C (1013 hPa)
Punto di fusione: 174 °C
MDL Number: MFCD00005788
CAS Number: 94-45-1
EINECS: 202-333-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68dfbe8a3b137577544e909026f43dfaa233f513&vid=68dfbe8a3b137577544e909026f43dfaa233f513&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR