Stampa…

2-Amino-1,3-propanediol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol

A1252-25G A1252-5G
TCIAA1252-25GEA 113 EUR
TCIAA1252-25G TCIAA1252-5G
2-Amino-1,3-propanediol ≥98.0% (by GC, titration analysis)
2-Amino-1,3-propanediol
Formula: C₃H₉NO₂
Peso molecolare: 91,11 g/mol
Punto di ebollizione: 116°C
Punto di fusione: 91…95 °C
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbae4806cad4331d67fd8deb1dfdb7423087ac2b&vid=dbae4806cad4331d67fd8deb1dfdb7423087ac2b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR