Methyl-2,4-dihydroxybenzoate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₈O₄ Punto di ebollizione: 300 °C (1013 hPa) Punto di fusione: 214…215 °C |
MDL Number:
MFCD00002276 CAS Number: 2150-47-2 EINECS: 218-428-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...