Stampa…

5-Iodouracil ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Iodopyrimidine-2,4(1H,3H)-dione, Iodouracil

I0219-25G I0219-5G
TCIAI0219-25GEA 119 EUR
TCIAI0219-25G TCIAI0219-5G
5-Iodouracil ≥99.0% (by HPLC, titration analysis)
5-Iodouracil
Formula: C₄H₃IN₂O₂
Peso molecolare: 237,98 g/mol
Punto di ebollizione: 401 °C (1013 hPa)
Punto di fusione: 270…274 °C
MDL Number: MFCD00006020
CAS Number: 696-07-1
EINECS: 211-788-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b7efbcb09236f3ad4e9508cee54ab4cbd05347&vid=a6b7efbcb09236f3ad4e9508cee54ab4cbd05347&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in dilute NH3aq. very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR