5-(Trifluoromethyl)uracil ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Trifluorothymine, 5-(Trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
T1622-1G
T1622-5G
TCIAT1622-1GEA
50
EUR
TCIAT1622-1G
TCIAT1622-5G
5-(Trifluoromethyl)uracil ≥97.0% (by HPLC)
5-(Trifluoromethyl)uracil
Formula:
C₅H₃F₃N₂O₂ Peso molecolare: 180,09 g/mol Punto di ebollizione: 350 °C (1013 hPa) Punto di fusione: 251…252 °C |
MDL Number:
MFCD00006024 CAS Number: 54-20-6 EINECS: 200-197-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | 97.0 to 102.0 % |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...