Stampa…

3,3'-Dinitrobenzophenone ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Bis(3-nitrophenyl) ketone, Bis(3-nitrophenyl)methanone

D1688-25G D1688-5G
TCIAD1688-25GEA 890 EUR
TCIAD1688-25G TCIAD1688-5G
3,3'-Dinitrobenzophenone ≥99.0% (by GC)
3,3'-Dinitrobenzophenone
Formula: C₁₃H₈N₂O₅
Peso molecolare: 272,22 g/mol
MDL Number: MFCD00039740
CAS Number: 21222-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afe8482d563e7e8edffc8d0d70d822b5642a9350&vid=afe8482d563e7e8edffc8d0d70d822b5642a9350&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 154.0 to 157.0 deg-C
Solubility in hot Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR