N-Hydroxyphthalimide ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxyisoindoline-1,3-dione, 2-Hydroxy-1H-isoindole-1,3(2H)-dione, N-Hydroxypthalimide
H0395-25G
H0395-500G
H0395-100G
TCIAH0395-25GEA
16
EUR
TCIAH0395-25G
TCIAH0395-500G
TCIAH0395-100G
N-Hydroxyphthalimide ≥99.0% (by titrimetric analysis)
N-Hydroxyphthalimide
Formula:
C₈H₅NO₃ Peso molecolare: 163,13 g/mol Punto di ebollizione: 412 °C (1013 hPa) Punto di fusione: 231°C Densità: 1,58 g/cm³ (20 °C) |
MDL Number:
MFCD00005891 CAS Number: 524-38-9 EINECS: 208-358-1 Merck Index: 13,04864 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...