Stampa…

1-Hexyl-3-methylimidazolium chloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 1-n-Hexyl-3-methylimidazolium chloride

H1097-25G H1097-5G
TCIAH1097-25GEA 75 EUR
TCIAH1097-25G TCIAH1097-5G
1-Hexyl-3-methylimidazolium chloride ≥98.0% (by titrimetric analysis)
1-Hexyl-3-methylimidazolium chloride
Formula: C₁₀H₁₉ClN₂
Peso molecolare: 202,73 g/mol
Punto di fusione: –85 °C
Densità: 1,05 g/cm³ (20 °C)
MDL Number: MFCD03093289
CAS Number: 171058-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84528dffd746794451e2c3ae70e6178b02476dac&vid=84528dffd746794451e2c3ae70e6178b02476dac&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow clear liquid
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR