1-Hexyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
H1227-25G
H1227-5G
TCIAH1227-25GEA
286
EUR
TCIAH1227-25G
TCIAH1227-5G
1-Hexyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
1-Hexyl-3-methylimidazolium bromide
Formula:
C₁₀H₁₉BrN₂ Punto di ebollizione: 395°C |
MDL Number:
MFCD03427612 CAS Number: 85100-78-3 |
|
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid to slightly cloudy liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...