Stampa…

3-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Methyl-3-chloroaniline

C1891-25G C1891-5G
TCIAC1891-25GEA 365 EUR
TCIAC1891-25G TCIAC1891-5G
3-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)
3-Chloro-N-methylaniline
Formula: C₇H₈ClN
Peso molecolare: 141,6 g/mol
Punto di ebollizione: 235°C
Densità: 1,174 g/cm³ (11.5 °C)
MDL Number: MFCD00052014
CAS Number: 7006-52-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b583a963a788cb08e7e30d4b25b117624eca68&vid=b3b583a963a788cb08e7e30d4b25b117624eca68&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR