1-Butyl-3-methylimidazolium iodide ≥96.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₁₅IN₂ Densità: 1,49 g/cm³ (20 °C) Flash Pt: 223 °C |
MDL Number:
MFCD08276307 CAS Number: 65039-05-6 |
|
Risultati del test di specificazione
Appearance | Orange to Brown to Dark red clear liquid |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...