Stampa…

1-Butyl-2,3-dimethylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-Butyl-2,3-dimethyl-3-imidazolium chloride

B2473-25G B2473-5G
TCIAB2473-25GEA 114 EUR
TCIAB2473-25G TCIAB2473-5G
1-Butyl-2,3-dimethylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Butyl-2,3-dimethylimidazolium chloride
Formula: C₉H₁₇ClN₂
Peso molecolare: 188,7 g/mol
MDL Number: MFCD03093291
CAS Number: 98892-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b96e991a036ebe2b44386594364bacebad5dd3f1&vid=b96e991a036ebe2b44386594364bacebad5dd3f1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR