Stampa…

4-Bromo-o-phenylenediamine ≥96.0% (by GC)

Fornitore: TCI
Attenzione

B3181-25G B3181-5G
TCIAB3181-25GEA 126 EUR
TCIAB3181-25G TCIAB3181-5G
4-Bromo-o-phenylenediamine ≥96.0% (by GC)
4-Bromo-o-phenylenediamine
Formula: C₆H₇BrN₂
MDL Number: MFCD02660622
CAS Number: 1575-37-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8fa446484cea2ea6b0dd4910d4ddc73ff29b4c4&vid=d8fa446484cea2ea6b0dd4910d4ddc73ff29b4c4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR