Stampa…

2-(4-Aminophenyl)ethanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Aminophenethyl alcohol

A0394-25G A0394-100G
TCIAA0394-25GEA 66 EUR
TCIAA0394-25G TCIAA0394-100G
2-(4-Aminophenyl)ethanol ≥98.0% (by GC, titration analysis)
2-(4-Aminophenyl)ethanol
Formula: C₈H₁₁NO
Peso molecolare: 137,18 g/mol
Punto di ebollizione: 255°C
Punto di fusione: 108 °C
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d5a4180fdb311289185de1de1e8f4b47cd04e9&vid=92d5a4180fdb311289185de1de1e8f4b47cd04e9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 110.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR