Stampa…
Adipoile dicloruro ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Pericolo
Synonyms: Adipoyl chloride
A0169-100G A0169-25G A0169-500G
TCIAA0169-100GEA 102 EUR
TCIAA0169-100G TCIAA0169-25G TCIAA0169-500G
Adipoile dicloruro ≥98.0% (by GC, titration analysis)
Adipoile dicloruro
Formula: C₆H₈Cl₂O₂
Peso molecolare: 183,03 g/mol
Punto di ebollizione: 115°C
Densità: 1,2563 g/cm³ (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47fb97071e9a71049cab00f389f93e76d80dc162&vid=47fb97071e9a71049cab00f389f93e76d80dc162&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %