2,6-Dicloro-4-nitroanisolo ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
D2872-1G
D2872-5G
TCIAD2872-1GEA
46
EUR
TCIAD2872-1G
TCIAD2872-5G
2,6-Dicloro-4-nitroanisolo ≥98.0% (by GC)
2,6-Dicloro-4-nitroanisolo
Formula:
C₇H₅Cl₂NO₃ Peso molecolare: 222,03 g/mol Punto di fusione: 102 °C |
MDL Number:
MFCD00061129 CAS Number: 17742-69-7 EINECS: 403-350-6 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...