Stampa…

2,4,6-Trimethyl-m-phenylenediamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3-Diamino-2,4,6-trimethylbenzene, 2,4-Diamino-1,3,5-trimethylbenzene, 2,4,6-Trimethyl-1,3-phenylenediamine, 2,4-Diaminomesitylene, 2,4,6-Trimethylbenzene-1,3-diamine

T1275-25G T1275-100G
TCIAT1275-25GEA 85 EUR
TCIAT1275-25G TCIAT1275-100G
2,4,6-Trimethyl-m-phenylenediamine ≥98.0% (by GC)
2,4,6-Trimethyl-m-phenylenediamine
Formula: C₉H₁₄N₂
Peso molecolare: 150,22 g/mol
MDL Number: MFCD00010149
CAS Number: 3102-70-3
EINECS: 221-456-9
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cf84f78f9f41d7cb3b511d384c024b65edd9fae&vid=7cf84f78f9f41d7cb3b511d384c024b65edd9fae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 89.0 to 93.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR