Stampa…

Isophthalaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde

I0153-25G I0153-250G
TCIAI0153-25GEA 32 EUR
TCIAI0153-25G TCIAI0153-250G
Isophthalaldehyde ≥98.0% (by GC)
Isophthalaldehyde
Formula: C₈H₆O₂
Peso molecolare: 134,13 g/mol
Punto di ebollizione: 136°C
Punto di fusione: 89…91 °C
MDL Number: MFCD00003372
CAS Number: 626-19-7
EINECS: 210-935-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52fdc705d07e69e4865a1ffc856d3004de7abf7&vid=d52fdc705d07e69e4865a1ffc856d3004de7abf7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR