Stampa…

Ftalimide ≥98.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: Phthalic acid imide

P0402-25G P0402-500G
TCIAP0402-25GEA 16 EUR
TCIAP0402-25G TCIAP0402-500G
Ftalimide ≥98.0% (by GC, titration analysis)
Ftalimide
Formula: C₈H₅NO₂
Peso molecolare: 147.13 g/mol
Punto di ebollizione: 366
Flash Pt: 165
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005881
CAS Number: 85-41-6
EINECS: 201-603-3
Merck Index: 14,07373
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40eeeeca3251e67c3a81f1484d60e083ece988a&vid=d40eeeeca3251e67c3a81f1484d60e083ece988a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 230.0 to 238.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR