4-Methylbenzyl mercaptan ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
p-Xylene-α-thiol
M0682-25ML
M0682-5ML
TCIAM0682-25MLEA
385
EUR
TCIAM0682-25ML
TCIAM0682-5ML
4-Methylbenzyl mercaptan ≥97.0% (by GC)
4-Methylbenzyl mercaptan
Formula:
C₈H₁₀S Peso molecolare: 138,23 g/mol Punto di ebollizione: 118°C Densità: 1,034 g/cm³ (23 °C) |
MDL Number:
MFCD00039652 CAS Number: 4498-99-1 EINECS: 224-796-6 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...