1-Adamantanemethanol ≥99.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₈O Punto di ebollizione: 253 °C (1013 hPa) Punto di fusione: 115…118 °C |
MDL Number:
MFCD00074751 CAS Number: 770-71-8 EINECS: 212-225-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 115.0 to 119.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...