5-Fluoro-2-methylbenzyl bromide ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-(Bromomethyl)-4-fluoro-1-methylbenzene
F0748-1G
F0748-5G
TCIAF0748-1GEA
65
EUR
TCIAF0748-1G
TCIAF0748-5G
5-Fluoro-2-methylbenzyl bromide ≥97.0% (by GC)
5-Fluoro-2-methylbenzyl bromide
Formula:
C₈H₈BrF Peso molecolare: 203,05 g/mol |
MDL Number:
MFCD01631528 CAS Number: 261951-71-7 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...