Stampa…

N-(4-Chlorophenyl)phthalimide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(4-Chlorophenyl)-1H-isoindole-1,3(2H)-dione

C1958-5G C1958-25G
TCIAC1958-5GEA 73 EUR
TCIAC1958-5G TCIAC1958-25G
N-(4-Chlorophenyl)phthalimide ≥98.0% (by GC)
N-(4-Chlorophenyl)phthalimide
Formula: C₁₄H₈ClNO₂
Peso molecolare: 257,68 g/mol
MDL Number: MFCD00023048
CAS Number: 7386-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94392e75f2ed1c37e0f92bcb9fd3a91d84e16a69&vid=94392e75f2ed1c37e0f92bcb9fd3a91d84e16a69&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 196.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR