Stampa…

N-(2-Hydroxyethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI
Attenzione

Synonyms: 2-(2-Hydroxyethyl)isoindoline-1,3-dione

H0245-25G H0245-500G
TCIAH0245-25GEA 29 EUR
TCIAH0245-25G TCIAH0245-500G
N-(2-Hydroxyethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(2-Hydroxyethyl)phthalimide
Formula: C₁₀H₉NO₃
MDL Number: MFCD00005903
CAS Number: 3891-07-4
EINECS: 223-434-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fade761423803bc7cf6f0dec7e34ceb24b4f5a13&vid=fade761423803bc7cf6f0dec7e34ceb24b4f5a13&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 128.0 to 131.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR