Stampa…

N-(2-Bromoethyl)phthalimide ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-(2-Bromoethyl)isoindoline-1,3-dione

B0597-25G B0597-100G
TCIAB0597-25GEA 33 EUR
TCIAB0597-25G TCIAB0597-100G
N-(2-Bromoethyl)phthalimide ≥98.0% (by GC, titration analysis)
N-(2-Bromoethyl)phthalimide
Formula: C₁₀H₈BrNO₂
Peso molecolare: 254,08 g/mol
Punto di ebollizione: 318°C
Punto di fusione: 79…82 °C
MDL Number: MFCD00005902
CAS Number: 574-98-1
EINECS: 209-379-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7142673f15e74bc6496cd514f10cc01b5033565d&vid=7142673f15e74bc6496cd514f10cc01b5033565d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR