Stampa…

Benzyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium bromide

B2980-25G B2980-250G
TCIAB2980-25GEA 28 EUR
TCIAB2980-25G TCIAB2980-250G
Benzyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Benzyltrimethylammonium bromide
Formula: C₁₀H₁₆BrN
Peso molecolare: 230.15 g/mol
Punto di fusione: 235
MDL Number: MFCD00011780
CAS Number: 5350-41-4
EINECS: 226-320-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7a9af96c67582cac230fdc0b695be52c5587b0f&vid=e7a9af96c67582cac230fdc0b695be52c5587b0f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR