Stampa…

Ethylurea ≥95.0% (by titrimetric analysis)

Fornitore: TCI

E0192-25G E0192-500G
TCIAE0192-25GEA 35 EUR
TCIAE0192-25G TCIAE0192-500G
Ethylurea ≥95.0% (by titrimetric analysis)
Ethylurea
Formula: C₃H₈N₂O
Punto di ebollizione: 258 °C (1013 hPa)
Punto di fusione: 94,55 °C
Densità: 1,213 g/cm³ (18 °C)
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1911f70a1dfacc9fa3597ac024be70259189fe20&vid=1911f70a1dfacc9fa3597ac024be70259189fe20&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 91.0 to 96.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR