Ethyl-p-tolylacetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl p-Tolylacetate, (4-Methylphenyl)acetic acid ethyl ester, p-Tolylacetic acid ethyl ester
T1227-25G
T1227-5G
TCIAT1227-25GEA
192
EUR
TCIAT1227-25G
TCIAT1227-5G
Ethyl-p-tolylacetate ≥98.0% (by GC)
Ethyl-p-tolylacetate
Formula:
(C₆H₄(CH₃))CH₂COOCH₂CH₃ Peso molecolare: 178,23 g/mol Punto di ebollizione: 240°C Densità: 1 g/cm³ (20 °C) Flash Pt: 109°C |
MDL Number:
MFCD00009181 CAS Number: 14062-19-2 EINECS: 237-904-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...