Stampa…

(4-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

A2182-25G A2182-5G
TCIAA2182-25GEA 101 EUR
TCIAA2182-25G TCIAA2182-5G
(4-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(4-Acetylphenoxy)acetic acid
Formula: (C₆H₄(C(O)CH₃))OCH₂COOH
MDL Number: MFCD00014363
CAS Number: 1878-81-5
EINECS: 217-524-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8ce456935597c4865dc80fa6fc9001bb2a8aab&vid=ea8ce456935597c4865dc80fa6fc9001bb2a8aab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR