1-Bromo-3-phenoxypropane ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-Bromopropyl phenyl ether, (3-Bromopropoxy)benzene, 3-Phenoxypropyl bromide
P1027-25G
P1027-250G
TCIAP1027-25GEA
56
EUR
TCIAP1027-25G
TCIAP1027-250G
1-Bromo-3-phenoxypropane ≥95.0% (by GC)
1-Bromo-3-phenoxypropane
Formula:
(C₆H₅)OCH₂CH₂CH₂Br Peso molecolare: 215,09 g/mol Punto di ebollizione: 138°C Densità: 1,365 g/cm³ (16 °C) Flash Pt: 96°C |
MDL Number:
MFCD00000256 CAS Number: 588-63-6 EINECS: 209-623-4 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...